missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCRN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121853
This item is not returnable.
View return policy
Description
SCRN1 Polyclonal Antibody for Western Blot, IHC (P)
This gene likely encodes a member of the secernin family of proteins. A similar protein in rat functions in regulation of exocytosis in mast cells. Alternatively spliced transcript variants have been described.
Specifications
| SCRN1 | |
| Polyclonal | |
| Unconjugated | |
| SCRN1 | |
| 2810019K23Rik; 6330535A03Rik; AI852905; Kiaa0193; mKIAA0193; Scrn1; secernin 1; secernin-1; SES1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 69938, 9805 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q12765, Q9CZC8 | |
| SCRN1 | |
| Recombinant protein Secernin 1. The antigen corresponds to amino acid range 2-414 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction