missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Screw Caps for Polystyrene Roller Bottles

Click to view available options
Thread Size (Metric):
83 mm
Unit Size:
Pack of 100
Description
- Sterile
- Non-cytotoxic
- Non-pyrogenic
- Free of detectable DNase, RNase, human DNA
Specifications
Specifications
| Color | Blue |
| No. per Case | 100 |
| Sterility | Sterile |
| Thread Size (Metric) | 83 mm |
| Neck Finish | 83 mm |
| Product Type | Screw Cap |
| For Use With (Equipment) | Polystyrene roller bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction