Learn More
BOLA Screw Caps
Product Code. p-200110014
Click to view available options
Thread Size (Metric):
GL 14
GL 18
GL 25
GL 32
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 10466591
Brand: BOLA H98601
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Screw Caps PTFE
Screw cap with handy knurl, suitable for GL threads, made of PTFE. After assembly, the product is only exposed to PTFE.
Specifications
Color White
Sterility Non-sterile
Thread Size (Metric) GL 14
Neck Finish 98 mm
Certifications/Compliance FDA
Disposable Reusable
Autoclavable Autoclavable