missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Witeg™ Screw Cap for Multiple Distributors
Click to view available options
Thread Size (Metric):
GL 80
GL 100
Unit Size:
Each
Description
- Grey in color
- Accessory item
- Polypropylene construction
- Available for thread GL 80 or GL 100
- Provided 10 per pack
Specifications
Specifications
| Color | Gray |
| Material | Polypropylene |
| Quantity | 10 Pk. |
| Thread Size (Metric) | GL 80 |
| Product Type | Screw Cap |
| For Use With (Equipment) | Multiple distributors |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction