Learn More
DWK Life Sciences Screw Cap
Screw caps provide a tight seal without the need for crimping and decapping tools.
15.00 € - 50.70 €
Specifications
For Use With (Application) | High Performance Liquid Chromatography (HPLC) |
---|---|
Hardness | 45 ± 5 shore A |
Autoclavable | Autoclavable |
Quantity | 100 |
Product Code | Brand | Type | Color | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Color | Material | Price | Quantity & Availability | |||
16676882
|
DWK Life Sciences
225784 |
Open Top Cap | Black | White Silicone, Red PTFE, Polypropylene |
28.45 €
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
16686882
|
DWK Life Sciences
225785 |
Open Top Cap | Black | White Silicone, Red PTFE, Pre-slit (single), Polypropylene |
50.70 €
Pack of 100 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||
16696882
|
DWK Life Sciences
225786 |
Open Top Cap | Black | Red PTFE, White Silicone, Red PTFE, Polypropylene |
37.85 €
Pack of 100 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||
16626892
|
DWK Life Sciences
225789 |
Open Top Cap | Blue | Red PTFE, White Silicone, Red PTFE, Polypropylene |
34.85 €
Pack of 100 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
16646892
|
DWK Life Sciences
225791 |
Open Top Cap | White | Red Rubber, Clear PTFE, Polypropylene |
15.00 €
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
16656892
|
DWK Life Sciences
225792 |
Open Top Cap | White | White Silicone, Red PTFE, Polypropylene |
23.45 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
16666892
|
DWK Life Sciences
225793 |
Open Top Cap | White | Red PTFE, White Silicone, Red PTFE, Polypropylene |
34.85 €
Pack of 100 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||
16606902
|
DWK Life Sciences
225797 |
Open Top Cap | Black | White Silicone, Red PTFE, Polypropylene |
34.25 €
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
Description
- Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability
Specifications
High Performance Liquid Chromatography (HPLC) | |
45 ± 5 shore A | |
Autoclavable | |
100 |