missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP3/SYCP3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3750-SP
This item is not returnable.
View return policy
Description
SCP3/SYCP3 Polyclonal specifically detects SCP3/SYCP3 in Human samples. It is validated for Western Blot.Specifications
| SCP3/SYCP3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COR1, MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, SYCP3, synaptonemal complex protein 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 50511 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q8IZU3 | |
| SYCP3 | |
| E. coli-derived recombinant human SCP3/SYCP3 Met1-Phe236 Accession # Q8IZU3 | |
| 25 μg | |
| Primary | |
| Detects human SCP3/SYCP3 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?