missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCNM1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15928594
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15928594 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15928594 Supplier Invitrogen™ Supplier No. PA565860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SREPEPAAGPQ AEESATVSAP APMSPTRRRA LDHYLTLRSS GWIPDGRGRW VKDENVEFDS DEEEPPDLP Highest antigen sequence identity to the following orthologs - mouse 41%, rat 76%.

SCNM1 is a zinc finger protein and putative splicing factor. In mice, Scnm1 modifies phenotypic expression of Scn8a mutations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCNM1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCNM1
Gene Accession No. Q9BWG6
Gene Alias 3110001I17Rik; Scnm1; Scnm1-ps; Sodium channel modifier 1; sodium channel modifier 1, pseudogene
Gene Symbols SCNM1
Host Species Rabbit
Immunogen Recombinant Human SCNM1. Recombinant protein control fragment (Product #RP-106528).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79005
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.