missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scn2a1 Mouse anti-Mouse, Rat, RPE, Clone:CJ6, Abnova™
Mouse monoclonal antibody raised against partial recombinant rat Scn2a1.
Brand: Abnova MAB18028.100ug
This item is not returnable.
View return policy
Specifications
| Scn2a1 | |
| Monoclonal | |
| R-PE | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Scn2a1 | |
| Mouse | |
| 100 μg | |
| Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| CJ6 | |
| sodium channel, voltage-gated, type II, alpha 1 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| NachII, Nav1.2, RII/RIIA, RNSCPIIR, SCN, SCN2A, Scn2a2, ScpII | |
| Recombinant protein corresponding to amino acids 1882-2005 at C-terminus of rat Scn2a1. | |
| 24766 | |
| Store at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction