missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCL/Tal1 Antibody (1E6), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50285C
This item is not returnable.
View return policy
Description
SCL/Tal1 Monoclonal specifically detects SCL/Tal1 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| SCL/Tal1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TAL1 | |
| GST-SCL/Tal1 (N-terminal) fusion protein | |
| 0.1 mL | |
| Primary | |
| Zebrafish | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1000000 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6886 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction