Learn More
SI Analytiks™ ScienceLine Ion Selective Electrode
SI Analytics ScienceLine Ion Selective Electrodes possess the advantage of a much lower temperature sensitivity and a metal ion free reference system.
Brand:  SI Analytiks™ 285216313
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 10051092
  • 870.00 € / Each
Estimated Shipment: 17-08-2022
to see stock.
Add to basket
Description
Description
  • ScienceLine sensors for ammonia, sodium, oxygen, ion-selective indicator electrodes
  • Also measure pH and temperature
Specifications
Specifications
Fluoride
F 1100 A
0°C to 80°C
0.02 mg/L to Saturated
5 to 7
Selective
Electrode