Learn More
SCH900776, MedChemExpress
Product Code. 16499167
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16499167
Brand: Medchem Express HY15532
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress SCH900776 (MK-8776) is a potent, selective and orally bioavailable inhibitor of checkpoint kinase1 (Chk1) with an IC50 of 3 nM. SCH900776 shows 50- and 500-fold selectivity over CDK2 and Chk2, respectively.
SCH900776 (MK-8776) is a potent, selective and orally bioavailable inhibitor of checkpoint kinase1 (Chk1) with an IC50 of 3 nM. SCH900776 shows 50- and 500-fold selectivity over CDK2 and Chk2, respectively.
Specifications
Chemical Name or Material SCH900776
Physical Form Solid
CAS 891494-63-6
Color Earth Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C15H18BrN7
For Use With (Application) Cancer-Kinase/protease
Synonym MK-8776
Solubility Information DMSO : ≥ 100 mg/mL (265.78 mM)
SMILES NC1=C(Br)C([C@H]2CNCCC2)=NC3=C(C4=CN(C)N=C4)C=NN13
Molecular Weight (g/mol) 376.25
Formula Weight 376.25
Percent Purity 98.71%
Grade Research