Learn More
Scaffold attachment factor B2 Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Scaffold attachment factor B2 Polyclonal antibody specifically detects Scaffold attachment factor B2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Scaffold attachment factor B2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | KIAA0138SAF-B2, scaffold attachment factor B2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-255 of human Scaffold attachment factor B2 (NP_055464.1).,, Sequence:, MAETLPGSGDSGPGTASLGPGVAETGTRRLSELRVIDLRAELKKRNLDTGGNKSVLMERLKKAVKEEGQDPDEIGIELEATSKKSAKRCVKGLKMEEEGTEDNGLEDDSRDGQEDMEASLENLQNMGMMDMSVLDETEVANSSAPDFGEDGTDGLLDSFCDSKEYVAAQLRQLPAQPPEHAVDGEGFKNTLETSSLNFKVTPDIEESLLEPENEKILDILGETCKSEPVKEESSELEQPFAQDTSSVGPDRKLAE |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |