missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sca1 (Ly-6A/E) Rat, anti-Mouse, FITC, Clone: E13-161.7, STEMCELL Technologies
Description
- The E13-161.7 antibody reacts with Sca1 (Stem Cell Antigen-1 or Ly-6A/E), an 18 kDa GPI-linked protein belonging to the lymphocyte activation protein-6 (Ly-6) family
- Sca1 is expressed on the surface of hematopoietic stem and progenitor cells, myeloid cells, and peripheral B and T lymphocytes
- Sca1 expression levels are strongly upregulated in all strains upon cellular activation
- Sca1 is involved in the regulation of T and B cell responses and is believed to play roles in the differentiation, proliferation, and survival of a variety of stem cells
- Sca1 has emerged as a phenotypic marker of choice for identifying and isolating hematopoietic stem and progenitor cells
Specifications
Specifications
| Antigen | Sca1 (Ly-6A/E) |
| Applications | Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Clone | E13-161.7 |
| Conjugate | FITC |
| Host Species | Rat |
| Quantity | 50 μg |
| Primary or Secondary | Primary |
| Target Species | Mouse |
| Isotype | IgG2a κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?