missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SC 144 hydrochloride

Description
SC 144 hydrochloride is an inhibitor of gp130 (IC50 = 0.72 μM); blocks cytokine-triggered gp130 signaling. SC 144 hydrochloride induces gp130 phosphorylation and down regulates gp130 glycosylation; also abrogates STAT3 phosphorylation, nuclear translocation and inhibits the expression of downstream target genes. SC 144 hydrochloride delays tumor growth in a mouse xenograft model of human ovarian cancer. Orally active.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | SC 144 hydrochloride |
| Purity | >99% |
| Molecular Weight (g/mol) | 358.76 |
| Inhibitors | SC 144 hydrochloride |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?