missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies SB431542 (Hydrate)
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- SB431542 is a selective and potent inhibitor of the TGF-β/Activin/NODAL pathway that inhibits ALK5 (IC50 = 94 nM), ALK4 (IC50 = 140 nM), and ALK7 by competing for the ATP binding site
- It does not inhibit the BMP type I receptors ALK2, ALK3, and ALK6
- This product is supplied as the hydrate form of the molecule
Specifications
Specifications
| Synonym | SB-431542 |
| Quantity | 1 mg |
| Molecular Formula | C22H16N4O3 · XH2O |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction