missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies SB431542 (Hydrate)

Product Code. p-7217564
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17170042

Brand: STEMCELL Technologies 72234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activin/BMP/TGFβ pathway inhibitor; Inhibits ALK4, ALK5, and ALK7

  • SB431542 is a selective and potent inhibitor of the TGF-β/Activin/NODAL pathway that inhibits ALK5 (IC50 = 94 nM), ALK4 (IC50 = 140 nM), and ALK7 by competing for the ATP binding site
  • It does not inhibit the BMP type I receptors ALK2, ALK3, and ALK6
  • This product is supplied as the hydrate form of the molecule

Specifications

CAS 301836-41-9
Synonym SB-431542
Quantity 10 mg
Molecular Formula C22H16N4O3
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.