Learn More
STEMCELL Technologies SB431542 (Hydrate)
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- SB431542 is a selective and potent inhibitor of the TGF-β/Activin/NODAL pathway that inhibits ALK5 (IC50 = 94 nM), ALK4 (IC50 = 140 nM), and ALK7 by competing for the ATP binding site
- It does not inhibit the BMP type I receptors ALK2, ALK3, and ALK6
- This product is supplied as the hydrate form of the molecule
Specifications
Specifications
| CAS | 301836-41-9 |
| Synonym | SB-431542 |
| Quantity | 10 mg |
| Molecular Formula | C22H16N4O3 |
| Purity | ≥98% |