Learn More
SB 525334, MedChemExpress
Product Code. 16454948
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16454948
Brand: Medchem Express HY12043
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress SB 525334 is a potent and selective transforming growth factor β1 receptor (ALK5) inhibitor with an IC50 of 14.3 nM.
SB 525334 is a potent and selective transforming growth factor β1 receptor (ALK5) inhibitor with an IC50 of 14.3 nM.
Specifications
Chemical Name or Material SB 525334
CAS 356559-20-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H21N5
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (291.19 mM; Need ultrasonic)
SMILES CC(C)(C1=NC(C2=CC=CC(C)=N2)=C(C3=CC=C4N=CC=NC4=C3)N1)C
Molecular Weight (g/mol) 343.42
Formula Weight 343.42
Percent Purity 95.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease