Learn More
SB-222200, MedChemExpress
Product Code. 16476768
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16476768
Brand: Medchem Express HY1572210mg
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress SB-222200 is a potent, selective, orally active and blood-brain barrier (BBB) penetrant NK-3 receptor antagonist. SB-222200 is developed for central nervous system (CNS) disorders.
SB-222200 is a potent, selective, orally active and blood-brain barrier (BBB) penetrant NK-3 receptor antagonist. SB-222200 is developed for central nervous system (CNS) disorders.
Specifications
Chemical Name or Material SB-222200
CAS 174635-69-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H24N2O
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (262.83 mM)
SMILES O=C(C1=C(C)C(C2=CC=CC=C2)=NC3=CC=CC=C13)N[C@H](C4=CC=CC=C4)CC
Molecular Weight (g/mol) 380.48
Formula Weight 380.48
Percent Purity 98.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation