missing translation for 'onlineSavingsMsg'
Learn More

SB 216763, MedChemExpress

Product Code. p-200098215
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447667

Brand: Medchem Express HY12012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SB 216763 is potent, selective and ATP-competitive GSK-3 inhibitor with IC50s of 34.3 nM for both GSK-3α and GSK-3β.

SB 216763 is potent, selective and ATP-competitive GSK-3 inhibitor with IC50s of 34.3 nM for both GSK-3α and GSK-3β.

Specifications

Chemical Name or Material SB 216763
CAS 280744-09-4
Color Orange Red
Purity Grade Notes Research
Molecular Formula C19H12Cl2N2O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (269.38 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1=O)C(C2=CC=C(C=C2Cl)Cl)=C1C3=CN(C4=C3C=CC=C4)C
Molecular Weight (g/mol) 371.22
Formula Weight 371.22
Percent Purity 98.92%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.