Learn More
Fisherbrand™ Saturn Magnetic Stir Bars
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
40 mm
50 mm
Unit Size:
Each
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | FDA |
| Description | Spherical center with straight, cylindrical wings |
| For Use With (Equipment) | Flat or round-bottom vessels |
| Length (English) | 1.96 in. |
| Length (Metric) | 50 mm |
| Material | Magnet |
| Quantity | Each |
| Shape | Sphere, Cylinder |