Description
Quantitative Grade 393 Filter Papers
Made of 100% cotton linters. Ash-free (ash content ≤ 0.01 % according to DIN 540370). Wet-strengthened. Color-coded box for easy selection.
Specifications
Specifications
| Color Code | Purple |
| Format | Disc |
| Material | 100% Cotton Linters (>98%alpha-cellulose, Ash-free) |
| Particle Retention | 1 to 2 μm |
| Sterility | Non-sterile |
| Filtration Speed | 250 sec. |
| Grade | 393 |
| Quantity | 100/Pk. |
| Thickness (Metric) | 0.18 mm |
| Type | Quantitative Filter Paper |
| Show More |