Learn More
Sartorius™ Moisture Analyzer Labels
Product Code. 10357383 Shop All Sartorius Products
Change view
Click to view available options
Dimensions (LxW):
20 m x 57 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) unitSize
10357383 20 m x 57 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10357383 Supplier Sartorius™ Supplier No. 69Y03247
Only null left
Add to basket
Add to basket
Adhesive-backed, standard label paper in continuous roll
Order Info
L: 56 ft. (20m); W: 2.2 in. (57mm)
TRUSTED_SUSTAINABILITY
Specifications
Description Adhesive label, continuous roll
Color White
Dimensions (LxW) 20 m x 57 mm
Length (Metric) 20 m
Height (Metric) 57 mm