missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartopure® PP3 Midicaps® 5 µm Size 7

Brand:  Sartorius 5055342P7--OO--A

261.63 EUR valid until 2026-05-31
BEST PRICE on Promo! Use promo code "28017" to get your promotional price.


Sartopure® PP3 Midicaps® 5 µm Size 7

Product Code. 18894320

  • 275.40 € / Pack of 1
Optimize Your Filtration Workflow with Proven Performance
Use promo code 28017
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.