Learn More
Sartorius Stedim Biotech™ Sartobind Q Pico 0.08 mL
Strong ion exchangers for the polishing of monoclonal antibodies for the removal of viruses, DNA, host cell proteins, leached Protein A, and endotoxins.
Brand: Sartorius Stedim Biotech™ 92IEXQ42DD-11--D
389.81 EUR valid until 2026-05-31
BEST PRICE on Promo! Use promo code "28017" to get your promotional price.
Description
- Strong ion exchangers are perfect tools for the polishing of monoclonal antibodies for the removal of viruses, DNA, host cell proteins, leached Protein A, and endotoxins.
- As size exclusion effects are neglectible, they are suited for purification of viruses such as adeno or lentiviruses displaying one order of magnitude higher dynamic binding capacity.
Specifications
| Autoclavable | |
| Stabilized Reinforced Cellulose | |
| 2.9 cm2 | |
| Polypropylene, polyester | |
| 0.6 Mpa | |
| Membrane Adsorber, Ligand: Quaternary Ammonium |
| Conventional filter design | |
| 3 to 5 μm | |
| 0.8 to 2.4 mL/min. | |
| Female Luer Lock | |
| 10/pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.