missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike S1 Protein Antibody (1035206) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB105403C
This item is not returnable.
View return policy
Description
SARS-CoV-2 Spike S1 Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| SARS-CoV-2 Spike S1 Protein | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| SARS-CoV-2 | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 1035206 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| SARS-CoV-2 | |
| HEK293-derived SARS-CoV-2 Spike S1 Subunit protein, Accession # YP_009724390.1 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction