missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike RBD Antibody, R&D Systems™
Mouse Monoclonal Antibody
201.00 € - 461.00 €
Specifications
| Antigen | Spike RBD |
|---|---|
| Clone | 1035414 |
| Dilution | ELISA, Blockade of Receptor-ligand Interaction 50 ug/mL |
| Applications | ELISA, Blocking Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18191791
|
R&D Systems
MAB10580-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18192851
|
R&D Systems
MAB10580-100 |
100 μg |
461.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for ELISA, Blocking AssaySpecifications
| Spike RBD | |
| ELISA, Blockade of Receptor-ligand Interaction 50 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Human embryonic kidney cell HEK293-derived SARS-CoV-2 Spike RBD Protein, Ala319-Phe541, Accession # YP_009724390.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1035414 | |
| ELISA, Blocking Assay | |
| Unconjugated | |
| Mouse | |
| SARS-CoV-2 | |
| YP_009724390.1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title