missing translation for 'onlineSavingsMsg'
Learn More

Sardomozide dihydrochloride, MedChemExpress

Product Code. 16465838
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16465838

Brand: Medchem Express HY13746B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sardomozide dihydrochloride is an S-adenosylmethionine decarboxylase (SAMDC) inhibitor with an IC50 of 5 nM.

Sardomozide dihydrochloride is an S-adenosylmethionine decarboxylase (SAMDC) inhibitor with an IC50 of 5 nM.

Specifications

Chemical Name or Material Sardomozide dihydrochloride
CAS 138794-73-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H16Cl2N6
Quantity 10 mg
Synonym CGP 48664A
Solubility Information DMSO : 25 mg/mL (82.46 mM; Need ultrasonic) ∣H2O : 10 mg/mL (32.98 mM; Need ultrasonic)
SMILES N=C(C1=CC=CC2=C1CC/C2=N\NC(N)=N)N.[H]Cl.[H]Cl
Molecular Weight (g/mol) 303.19
Formula Weight 303.19
Percent Purity 97.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.