missing translation for 'onlineSavingsMsg'
Learn More

SAR7334 hydrochloride, MedChemExpress

Product Code. 16496748
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496748

Brand: Medchem Express HY15699A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAR7334 hydrochloride is a potent and specific TRPC6 inhibitor, inhibiting TRPC6 currents with IC50 of 7.9 nM.

SAR7334 hydrochloride is a potent and specific TRPC6 inhibitor, inhibiting TRPC6 currents with IC50 of 7.9 nM.

Specifications

Chemical Name or Material SAR7334 hydrochloride
CAS 1333207-63-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H24Cl3N3O
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (226.87 mM; Need ultrasonic) ∣H2O : 50 mg/mL (113.43 mM; Need ultrasonic)
SMILES N[C@@H](C1)CCCN1[C@@H]2CC3=CC=CC=C3[C@H]2OC4=CC=C(C#N)C=C4Cl.Cl.Cl
Molecular Weight (g/mol) 440.79
Formula Weight 440.79
Percent Purity 95.61%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.