Learn More
Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap
For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap are comprehensive thermal cycler compatible.
Brand: Greiner Bio-One 683275
Additional Details : Weight : 0.29000kg
Description
- Thin-walled for heat transfer during amplification
- Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
- Packaging labelled with product symbols for warehousing
- Used for sample handling
Specifications
Green | |
PCR | |
Polypropylene | |
PCR Tubes with Attached Flat Caps |
0.2 mL | |
Non-sterile | |
Attached Frosted Flat Cap |