Learn More
Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap
For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap are comprehensive thermal cycler compatible.
Brand:  Greiner Bio-One 682276
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 13045053
  • 110.00 € / Pack of 1000
Estimated Shipment: 25-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
Specifications
Specifications
Yellow
PCR
Polypropylene
PCR Tubes with Attached Flat Caps
0.5 mL
Non-sterile
Attached Frosted Flat Cap