Learn More
MedChemExpress Sapacitabine
Product Code. 16463097
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. 16463097
Brand: MedChemExpress HY16445
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Sapacitabine is an orally available nucleoside analog prodrug that is structurally related to cytarabine.
Specifications
Molecular Weight (g/mol) 490.64 g/mol
Product Type Reference compound
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 98.51%
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (67.93 mM∣Need ultrasonic)
For Use With (Application) Cancer-Kinase/protease
CAS 151823-14-2
Synonym CS682∣ CYC682