Description
- Stainless steel sand baths are available for irregular samples when there is no suitable block available
- Low temperature covers help to reduce the air flow for additional temperature stability
Specifications
Specifications
| Holds | 1 Block |
| Material | Stainless Steel |
| Type | Sand Bath |
| Includes | Thermometer Well |
| No. of Blocks | 1 |
| Size | 1 Block |