Learn More
Thermo Scientific™ Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Product Code. 15457904 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Sample Cone, Nickel
Skimmer Cone, Ni, Insert
For Use With (Equipment):
iCAP MX and Q/Qnova ICP-MS
iCAP Q/Qnova Series ICP-MS
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes For Use With (Equipment) Unit Size
15457904 Skimmer Cone, Ni, Insert iCAP Q/Qnova Series ICP-MS Each
17919660 Sample Cone, Nickel iCAP MX and Q/Qnova ICP-MS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15457904 Supplier Thermo Scientific™ Supplier No. 1311870
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ sample and skimmer cones for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova series ICP-MS systems are available to suit your analysis requirements and designed for durability, performance and ease of use.
Assure robust, uncomplicated trace elemental analysis for your laboratory with the Thermo Scientific iCAP MX and iCAP Q/Qnova series ICP-MS systems. Our easily changeable Ni and Pt sample and skimmer cones are available to suit your analysis requirements and designed for durability, performance and ease of use.
Select high-quality consumables for consistent performance. These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in either solid Ni construction or Pt tipped for even greater durability and specific analysis requirements.
  • Sample cones made from nickel and platinum
  • Skimmer cones made from nickel and platinum
  • Platinum, boron free skimmer cone (for semiconductor-related applications)
  • Skimmer cone inserts with different channel length to tailor instrument characteristics for each application
Specifications
Type Skimmer Cone
For Use With (Equipment) iCAP Q/Qnova Series ICP-MS
Includes Skimmer Cone, Ni, Insert