Learn More
Salvianolic Acid A, TRC
Specifications
Specifications
| Chemical Name or Material | Salvianolic acid A |
| CAS | 96574-01-5 |
| Molecular Formula | C26 H22 O10 |
| Quantity | 5 mg |
| Synonym | (αR)-α-[[(2E)-3-[2-[(1E)-2-(3,4-Dihydroxyphenyl)ethenyl]-3,4-dihydroxyphenyl]-1-oxo-2-propen-1-yl]oxy]-3,4-dihydroxybenzenepropanoic Acid, (+)-Salvianolic acid A, Dan Phenolic Acid A, Salvianolic Acid |
| InChI Formula | InChI=1S/C26H22O10/c27-18-7-2-14(11-21(18)30)1-6-17-16(4-9-20(29)25(17)33)5-10-24(32)36-23(26(34)35)13-15-3-8-19(28)22(31)12-15/h1-12,23,27-31,33H,13H2,(H,34,35)/b6-1+,10-5+/t23-/m1/s1 |
| SMILES | OC(=O)[C@@H](Cc1ccc(O)c(O)c1)OC(=O)\C=C\c2ccc(O)c(O)c2\C=C\c3ccc(O)c(O)c3 |
| IUPAC Name | (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-[2-[(E)-2-(3,4-dihydroxyphenyl)ethenyl]-3,4-dihydroxyphenyl]prop-2-enoyl]oxypropanoic acid |
| Molecular Weight (g/mol) | 494.45 |
| Formula Weight | 494.1213 |