Learn More
Salvianolic acid A, MedChemExpress
Product Code. 16461378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461378 5 mg 5mg
16443856 25 mg 25mg
16438948 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16461378 Supplier Medchem Express Supplier No. HYN0318
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Salvianolic acid A could protect the blood brain barrier through matrix metallopeptidase 9 (MMP-9) inhibition and anti-inflammation.
Salvianolic acid A could protect the blood brain barrier through matrix metallopeptidase 9 (MMP-9) inhibition and anti-inflammation.
Specifications
Chemical Name or Material Salvianolic acid A
CAS 96574-01-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H22O10
Quantity 5 mg
Solubility Information DMSO : 83.33 mg/mL (168.53 mM; Need ultrasonic)
SMILES OC1=CC=C(/C=C/C(O[C@@H](C(O)=O)CC2=CC(O)=C(O)C=C2)=O)C(/C=C/C3=CC(O)=C(O)C=C3)=C1O
Molecular Weight (g/mol) 494.45
Formula Weight 494.45
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation