Learn More
Invitrogen™ Salmonella typhimurium Monoclonal Antibody (F68C)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17443
Description
The MA1-7443 antibody reacts with Salmonella sp. in bacterial and infected tissue samples. Specific for Salmonella typhimurium. MA1-7443 has been successfully used in ELISA applications. The MA1-7443 antibody was raised against Salmonella preparation.
The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major surface antigens; the H or flagellar antigen (phase 1 and 2), the O or somatic antigen (part of the LPS moiety) and the Vi or capsular antigen (referred to as K in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram negative bacteria. This LPS is composed of an O polysaccharide (O antigen) an R core and the endotoxic inner Lipid A. Endotoxins evoke fever and can activate complement, kinin and clotting factors.
Specifications
| Salmonella typhimurium | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA | |
| F68C | |
| Unconjugated | |
| Salmonellae | |
| Salmonella preparation. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.