missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salinomycin sodium salt, MedChemExpress
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
Description
Salinomycin sodium salt (Salinomycin sodium), an antibiotic potassium ionophore, is a potent inhibitor of Wnt/β-catenin signaling. Salinomycin sodium salt (Salinomycin sodium) acts on the Wnt/Fzd/LRP complex, blocks Wnt-induced LRP6 phosphorylation, and causes degradation of the LRP6 protein. Salinomycin sodium salt (Salinomycin sodium) shows selective activity against human cancer stem cells.
Specifications
Specifications
| Chemical Name or Material | Salinomycin sodium salt |
| Physical Form | Solid |
| CAS | 55721-31-8 |
| Color | Light Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C42H69NaO11 |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | Salinomycin sodium Sodium salinomycin |
| Solubility Information | DMSO : 100 mg/mL (129.37 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction