missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAGE1 Monoclonal Antibody (5C1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524474
This item is not returnable.
View return policy
Description
Peptide Sequence: PLPSNALSTV LPGLAYLATA DMPAMSTRDQ HATIIHNLRE EKKDNSQPTP DNVLSAVTPE LINLAGAGIP PMSTRDQYAT VNHHVHEARM ENGQRKQDN.
This gene belongs to a class of genes that are activated in tumors. These genes are expressed in tumors of different histologic types but not in normal tissues, except for spermatogenic cells and, for some, placenta. The proteins encoded by these genes appear to be strictly tumor specific, and hence may be excellent sources of antigens for cancer immunotherapy. This gene is expressed in sarcomas.
Specifications
| SAGE1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9NXZ1 | |
| SAGE1 | |
| SAGE1 (NP_061136, 357 a.a. ~ 455 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 5C1 | |
| Unconjugated | |
| SAGE1 | |
| Cancer/testis antigen 14; CT14; putative tumor antigen; SAGE; SAGE1; Sarcoma antigen 1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 55511 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction