Learn More
SCAT™ Safety Waste Cap 'LISA', V3.0 Electronic Level Control Extension
Product Code. 17611795 Shop All SCAT Europe Products
Click to view available options
Type:
Electronic Control
Level Control
Material:
PTFE
PTFE, Conductive
Unit Size:
Each
Pack of 2
Pack of 3
Product Code. 17611795
Brand: SCAT™ 350121
Please to purchase this item. Need a web account? Register with us today!
With the extension Level Control, you can use your LISA to monitor the fill level of your disposal container at any time.
LISA is the first modular system for HPLC waste, providing occupational safety and environmental protection with proven SCAT™ technology and maximum flexibility. Simply add connections and components as required so you can collect solvents, acids and alkalis safely at any time.
  • Simply unscrew the blind plug of the LISA, insert the level control, and connect cable to the signal box
  • Permanently displays the fill level in your disposal container and alerts when reached as soon as it is full
  • Available in electrically conductive and non-electrically conductive versions
Specifications
For Use With (Equipment) SCAT™ Safety Waste Cap 'LISA', V3.0
Material PTFE, Conductive
For Use With (Application) HPLC Systems
Type Electronic Control