Learn More
asecos™ Q-Pegasus-90 Safety Storage Cabinet
For unrestricted storage of flammable hazardous materials in work areas.
Brand: asecos™ 3000204830015
This item is not returnable.
View return policy
Description
- Robust safety storage for industrial sector
- Tall cabinet with wing doors
- Automatic closing (TSA)
- One-hand operation for double-door cabinets (AGT)
- With cylinder locking and locking state indicator
- Integrated adjusting aids to compensate uneven floor
- Sheet steel powder coated RAL 7035
Warranty
10 years
Specifications
| EN 14470-1, DIN EN 14727, GS certified, CE compliant | |
| 615 mm | |
| 522 x 1050 x 1647 mm | |
| 1953 mm | |
| Q90.195.120.WDAC | |
| 1193 mm | |
| Green | |
| Safety Storage Cabinet |
| 4 x drawer (30L) | |
| Door: Reseda green RAL 6011; Sheet Steel RAL 7035 | |
| Wing | |
| 1647 mm | |
| Powder-coated textured | |
| 1050 mm | |
| Steel |