Learn More
Sacubitril Sodium Salt, TRC
Product Code. 30134745
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134745 1 g 1g
30132663 500 mg 500mg
17923545 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134745 Supplier Toronto Research Chemicals Supplier No. TRCS0809601G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 149690-05-1
Molecular Formula C24 H28 N O5 . Na
Molecular Weight (g/mol) 433.473
Synonym [1,1'-Biphenyl]-4-pentanoic acid, γ-[(3-carboxy-1-oxopropyl)amino]-α-methyl-, α-ethyl ester, sodium salt (1:1), (αR,γS)-, [1,1'-Biphenyl]-4-pentanoic acid, γ-[(3-carboxy-1-oxopropyl)amino]-α-methyl-, α-ethyl ester, monosodium salt, [S-(R*,S*)]- (9CI), Sacubitril sodium, Sacubitril sodium salt, (2R,4S)-5-(Biphenyl-4-yl)-4-[(3-carboxypropionyl)amino]-2-methylpentanoic acid ethyl ester sodium salt, [1,1'-Biphenyl]-4-pentanoic acid, γ-[(3-carboxy-1-oxopropyl)amino]-α-methyl-, α-ethyl ester, sodium salt (1:1), (αR,γS)-, [1,1'-Biphenyl]-4-pentanoic acid, γ-[(3-carboxy-1-oxopropyl)amino]-α-methyl-, α-ethyl ester, monosodium salt, [S-(R*,S*)]- (9CI), Sac
IUPAC Name sodium;4-[[(2S,4R)-5-ethoxy-4-methyl-5-oxo-1-(4-phenylphenyl)pentan-2-yl]amino]-4-oxobutanoate
SMILES [Na+].CCOC(=O)[C@H](C)C[C@@H](Cc1ccc(cc1)c2ccccc2)NC(=O)CCC(=O)[O-]
Specifications
Quantity 1 g
InChI Formula InChI=1S/C24H29NO5.Na/c1-3-30-24(29)17(2)15-21(25-22(26)13-14-23(27)28)16-18-9-11-20(12-10-18)19-7-5-4-6-8-19;/h4-12,17,21H,3,13-16H2,1-2H3,(H,25,26)(H,27,28);/q;+1/p-1/t17-,21+;/m1./s1
Formula Weight 433.187
Percent Purity >95%
Chemical Name or Material Sacubitril Sodium
Grade HPLC