missing translation for 'onlineSavingsMsg'
Learn More

S1P5/EDG-8 Antibody (282503), CoraFluor™ 1, Novus Biologicals™

Product Code. 30153177 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30153177 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30153177 Supplier Novus Biologicals Supplier No. FAB3964CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

S1P5/EDG-8 Monoclonal antibody specifically detects S1P5/EDG-8 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen S1P5/EDG-8
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 282503
Conjugate CoraFluor 1
Formulation PBS
Gene Alias Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human S1P5/EDG-8, Met1-Asp398, Accession # Q9H228
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 53637
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.