missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A9 Antibody (S100A9/1075), PE , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11559PE
This item is not returnable.
View return policy
Description
S100A9 Monoclonal antibody specifically detects S100A9 in Human, Rat samples. It is validated for Flow Cytometry
Specifications
| S100A9 | |
| Monoclonal | |
| PE | |
| CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B) | |
| Recombinant human S100A9 protein (Uniprot: P06702) | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity, Signal Transduction, Tyrosine Kinases | |
| 6280 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| S100A9/1075 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction