Learn More
(S)-Viloxazine-d5 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-Viloxazine-d5 Hydrochloride |
| CAS | 1246816-39-6 |
| Molecular Formula | C13H15D5ClNO3 |
| Quantity | 1 mg |
| Synonym | (S)-2-[((2-Ethoxy-d5)phenoxy)methyl]morpholine Hydrochloride, (S)-2-[(2-Ethoxy-d5)phenoxymethyl]tetrahydro-1,4-oxazine Hydrochloride |
| InChI Formula | InChI=1S/C13H19NO3.ClH/c1-2-15-12-5-3-4-6-13(12)17-10-11-9-14-7-8-16-11;/h3-6,11,14H,2,7-10H2,1H3;1H/t11-;/m0./s1/i1D3,2D2; |
| SMILES | Cl.[2H]C([2H])([2H])C([2H])([2H])Oc1ccccc1OC[C@@H]2CNCCO2 |
| IUPAC Name | (2S)-2-[[2-(1,1,2,2,2-pentadeuterioethoxy)phenoxy]methyl]morpholine;hydrochloride |
| Molecular Weight (g/mol) | 278.79 |
| Formula Weight | 278.1446 |