Learn More
(S,S)Dinitroso Ethambutol, TRC
Specifications
Specifications
| Chemical Name or Material | N,N'-Dinitrosoethambutol |
| CAS | 2792161-95-4 |
| Molecular Formula | C10 H22 N4 O4 |
| Quantity | 250 mg |
| Synonym | N,N'-Dinitrosoethambutol |
| InChI Formula | InChI=1S/C10H22N4O4/c1-3-9(7-15)13(11-17)5-6-14(12-18)10(4-2)8-16/h9-10,15-16H,3-8H2,1-2H3/t9-,10-/m0/s1 |
| SMILES | CC[C@@H](CO)N(CCN(N=O)[C@@H](CC)CO)N=O |
| IUPAC Name | N-[(2S)-1-hydroxybutan-2-yl]-N-[2-[[(2S)-1-hydroxybutan-2-yl]-nitrosoamino]ethyl]nitrous amide |
| Molecular Weight (g/mol) | 262.3061 |
| Formula Weight | 262.1641 |