missing translation for 'onlineSavingsMsg'
Learn More

(S)-Rasagiline, MedChemExpress

Product Code. p-200096723
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16415998

Brand: Medchem Express HY14200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-Rasagiline (TVP1022) is the relatively inactive S-enantiomer form of Rasagiline. Rasagiline is a highly potent selective irreversible MAO inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively.

(S)-Rasagiline (TVP1022) is the relatively inactive S-enantiomer form of Rasagiline. Rasagiline is a highly potent selective irreversible MAO inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively.

Specifications

Chemical Name or Material (S)-Rasagiline
CAS 185517-74-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H13N
Quantity 10 mg
Synonym TVP1022 S-PAI
Solubility Information DMSO : 100 mg/mL (583.98 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (583.98 mM; Need ultrasonic)
SMILES C#CCN[C@H]1CCC2=CC=CC=C21
Molecular Weight (g/mol) 171.24
Formula Weight 171.24
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.