missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress (S,R,S)-AHPC-PEG6-C4-Cl

Product Code. p-200104420
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490096 2 g 2g
16471566 500 mg 500mg
16431019 1 g 1g
16424916 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490096 Supplier MedChemExpress Supplier No. HY103606

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress (S,R,S)-AHPC-PEG6-C4-Cl is a conjugate of ligands for E3 and 25-atom-length linker.

The connector of linker is Halogen group. (S,R,S)-AHPC-PEG6-C4-Cl incorporates the (S,R,S)-AHPC based VHL ligand and 6-unit PEG linker. (S,R,S)-AHPC-PEG6-C4-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays.

Specifications

Molecular Weight (g/mol) 827.47 g/mol
CAS 1835705-59-3
Content And Storage Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Quantity 2 g
Synonym VH032-PEG6-C4-Cl ∣ VHL Ligand-Linker Conjugates 10 ∣ E3 ligase Ligand-Linker Conjugates 9
Solubility Ethanol : 100 mg/mL (120.85 mM; Need ultrasonic) ∣ DMSO : ≥ 100 mg/mL (120.85 mM)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.