missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress (S,R,S)-AHPC-PEG4-NH2

Product Code. p-200104418
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470096 2 g 2g
16451566 500 mg 500mg
16411019 1 g 1g
16404916 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470096 Supplier MedChemExpress Supplier No. HY103604A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (S,R,S)-AHPC-PEG4-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 4-unit PEG linker used in PROTAC technology.

Specifications

Molecular Weight (g/mol) 663.83 g/mol
CAS 2010159-57-4
Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Quantity 2 g
Synonym VH032-PEG4-NH2 ∣ VHL Ligand-Linker Conjugates 4 ∣ E3 ligase Ligand-Linker Conjugates 7 Free Base
Solubility DMSO : 170 mg/mL (256.09 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.