missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress (S,R,S)-AHPC-PEG2-NH2 hydrochloride
Specifications
Specifications
| Molecular Weight (g/mol) | 612.18 g/mol |
| CAS | 2097973-72-1 |
| Content And Storage | 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
| Shipping Condition | Room temperature |
| Quantity | 500 mg |
| Synonym | VH032-PEG2-NH2 hydrochloride ∣ VHL Ligand-Linker Conjugates 3 hydrochloride ∣ E3 ligase Ligand-Linker Conjugates 6 |
| Solubility | H2O : 100 mg/mL (163.35 mM; Need ultrasonic) ∣ DMSO : 100 mg/mL (163.35 mM; Need ultrasonic) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?